MAN458951433 Manchester Organics

1,2,5-Trichloro-3-iodo-benzene

  • MDL number MFCD01318175
  • Cas Number : 216393-66-7
  • Molecular Weight : 307.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,2,5-trichloro-3-iodobenzene
Inchi Code 1S/C6H2Cl3I/c7-3-1-4(8)6(9)5(10)2-3/h1-2H
Inchi Key YLLMVDOSVGVBJC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/G50929
Availability details for
Quantity
  • Availability
  • Add to Cart