MAN465534084 Manchester Organics

3-[tert-Butyl(dimethyl)silyl]oxycyclobutanol

  • MDL number MFCD23105930
  • Cas Number : 1408074-89-4
  • Molecular Weight : 202.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1s,3s)-3-((tert-butyldimethylsilyl)oxy)cyclobutan-1-ol
Inchi Code 1S/C10H22O2Si/c1-10(2,3)13(4,5)12-9-6-8(11)7-9/h8-9,11H,6-7H2,1-5H3/t8-,9+
Inchi Key ILGIKHCMCDNBSK-DTORHVGOSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/L50979
Availability details for
Quantity
  • Availability
  • Add to Cart