MAN465534085 Manchester Organics

3-[tert-Butyl(dimethyl)silyl]oxycyclobutanone

  • MDL number MFCD23105928
  • Cas Number : 929913-18-8
  • Molecular Weight : 200.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/M50980
Availability details for
Quantity
  • Availability
  • Add to Cart