MAN491534281 Manchester Organics

(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine hydrochloride

  • MDL number MFCD16621156
  • Cas Number : 220352-38-5
  • Molecular Weight : 205.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/D37816
Availability details for
Quantity
  • Availability
  • Add to Cart