MAN491534297 Manchester Organics

2-Methylaminoacetonitrile

  • MDL number MFCD00058964
  • Molecular Weight : 70.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W51080
Availability details for
Quantity
  • Availability
  • Add to Cart