MAN491534604 Manchester Organics

2-(5-Chloro-2-fluoro-phenyl)acetonitrile

  • MDL number MFCD01631438
  • Cas Number : 75279-54-8
  • Molecular Weight : 169.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/F51388
Availability details for
Quantity
  • Availability
  • Add to Cart