MAN491534938 Manchester Organics

Prop-2-enoyl prop-2-enoate

  • MDL number MFCD00048146
  • Molecular Weight : 126.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D51731
Availability details for
Quantity
  • Availability
  • Add to Cart