MAN491535158 Manchester Organics

4-Bromo-2-chloro-5-methyl-pyridine

  • MDL number MFCD11100650
  • Molecular Weight : 206.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/N51947
Availability details for
Quantity
  • Availability
  • Add to Cart