MAN497530821 Manchester Organics

(6-Methylpyridazin-3-yl)methanamine dihydrochloride

  • MDL number MFCD28155130
  • Cas Number : 1630907-25-3
  • Molecular Weight : 196.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-methylpyridazin-3-yl)methanamine dihydrochloride
Inchi Code 1S/C6H9N3.2ClH/c1-5-2-3-6(4-7)9-8-5;;/h2-3H,4,7H2,1H3;2*1H
Inchi Key DHHBGVAVHZOVNW-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/G52125
Availability details for
Quantity
  • Availability
  • Add to Cart