MAN509871261 Manchester Organics

4-Imidazoleacetic acid hydrochloride

  • MDL number MFCD00012698
  • Cas Number : 3251-69-2
  • Molecular Weight : 162.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/B22095
Availability details for
Quantity
  • Availability
  • Add to Cart