MAN509871297 Manchester Organics

1-Methyl-2-oxo-piperidine-4-carboxylic acid

  • MDL number MFCD09863345
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K52151
Availability details for
Quantity
  • Availability
  • Add to Cart