MAN509871345 Manchester Organics

Pyridazine-3,6-dicarboxylic acid

  • MDL number MFCD01318693
  • Molecular Weight : 168.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y96888
Availability details for
Quantity
  • Availability
  • Add to Cart