MAN509871550 Manchester Organics

2,2,6,6-Tetramethylpiperidin-4-one hydrate

  • MDL number MFCD00005975
  • Cas Number : 826-36-8
  • Molecular Weight : 173.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/K39892
Availability details for
Quantity
  • Availability
  • Add to Cart