MAN509871647 Manchester Organics

1-Methylpyrrolo[2,3-b]pyridine-3-carboxylic acid

  • MDL number MFCD15529044
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/Y52416
Availability details for
Quantity
  • Availability
  • Add to Cart