MAN509871724 Manchester Organics

Tert-butyl (3R,4R)-3-amino-4-hydroxy-pyrrolidine-1-carboxylate

  • MDL number MFCD06858457
  • Molecular Weight : 202.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y52508
Availability details for
Quantity
  • Availability
  • Add to Cart