MAN509871787 Manchester Organics

(2-Methylpyrimidin-5-yl)methanamine

  • MDL number MFCD10696906
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S52571
Availability details for
Quantity
  • Availability
  • Add to Cart