MAN514346731 Manchester Organics

2,4-Dimethylpentan-3-amine hydrochloride

  • MDL number MFCD00015041
  • Molecular Weight : 151.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W31507
Availability details for
Quantity
  • Availability
  • Add to Cart