MAN514346777 Manchester Organics

2-Chloro-4-methoxy-benzoic acid

  • MDL number MFCD00085943
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C52627
Availability details for
Quantity
  • Availability
  • Add to Cart