MAN514346861 Manchester Organics

(2-Chloro-6-isopropyl-3-pyridyl)boronic acid

  • MDL number MFCD08274472
  • Molecular Weight : 199.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin GB
Product Link https://www.manchesterorganics.com/E52698
Availability details for
Quantity
  • Availability
  • Add to Cart