MAN517331977 Manchester Organics

5H,6H,7H,8H-imidazo[1,2-a]pyrazine hydrochloride

  • MDL number MFCD09878589
  • Cas Number : 1187830-84-7
  • Molecular Weight : 159.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Country of Origin GB
Product Link https://www.manchesterorganics.com/N37181
Availability details for
Quantity
  • Availability
  • Add to Cart