MAN517332418 Manchester Organics

tert-Butyl N-[(1R,3S)-3-hydroxycyclopentyl]carbamate

  • MDL number MFCD11226176
  • Cas Number : 225641-84-9
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/X53220
Availability details for
Quantity
  • Availability
  • Add to Cart