MAN517332451 Manchester Organics

(E)-1,1,1-Trichloro-4-ethoxy-but-3-en-2-one

  • MDL number MFCD11520438
  • Molecular Weight : 217.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/F53251
Availability details for
Quantity
  • Availability
  • Add to Cart