MAN517332455 Manchester Organics

1,3-Dimethylindazol-6-amine

  • MDL number MFCD18432764
  • Cas Number : 221681-92-1
  • Molecular Weight : 161.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/K53255
Availability details for
Quantity
  • Availability
  • Add to Cart