MAN517332629 Manchester Organics

tert-Butyl (2S)-2-cyclopropylpiperazine-1-carboxylate

  • MDL number MFCD08686636
  • Cas Number : 1240586-17-7
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/H53414
Availability details for
Quantity
  • Availability
  • Add to Cart