MAN517332813 Manchester Organics

(2S)-2-Methoxypropanoic acid

  • MDL number MFCD01632589
  • Cas Number : 23953-00-6
  • Molecular Weight : 104.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/P53604
Availability details for
Quantity
  • Availability
  • Add to Cart