MAN517332822 Manchester Organics

tert-Butyl 2-formyl-6-azaspiro[3.3]heptane-6-carboxylate

  • MDL number MFCD24857302
  • Cas Number : 1440960-67-7
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 6-formyl-2-azaspiro[3.3]heptane-2-carboxylate
Inchi Code 1S/C12H19NO3/c1-11(2,3)16-10(15)13-7-12(8-13)4-9(5-12)6-14/h6,9H,4-5,7-8H2,1-3H3
Inchi Key QAIHVJYVIUKZAR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/A53614
Availability details for
Quantity
  • Availability
  • Add to Cart