MAN517333012 Manchester Organics

Diphenyl(vinyl)sulfonium; trifluoromethanesulfonate

  • MDL number MFCD22123284
  • Molecular Weight : 362.38712

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V53793

Safety Information

MSDS https://www.manchesterorganics.com/V53793
Availability details for
Quantity
  • Availability
  • Add to Cart