MAN517333226 Manchester Organics

5-Fluoro-2-methylpyrimidine-4,6-diol

  • MDL number MFCD11520459
  • Cas Number : 1598-63-6
  • Molecular Weight : 144.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/S53997
Availability details for
Quantity
  • Availability
  • Add to Cart