MAN517333229 Manchester Organics

2-Fluoropyrimidin-4-amine

  • MDL number MFCD00769380
  • Cas Number : 96548-91-3
  • Molecular Weight : 113.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-4-pyrimidinamine
Inchi Code 1S/C4H4FN3/c5-4-7-2-1-3(6)8-4/h1-2H,(H2,6,7,8)
Inchi Key SPWJNWKKNZWCPA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/V54000
Availability details for
Quantity
  • Availability
  • Add to Cart