MAN517333358 Manchester Organics

2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid, 1,1-dimethylethyl ester

  • MDL number MFCD06411671
  • Molecular Weight : 198.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T96356
Availability details for
Quantity
  • Availability
  • Add to Cart