MAN517333396 Manchester Organics

tert-Butyl N-[3,3-difluoro-1-(hydroxymethyl)cyclobutyl]carbamate

  • MDL number MFCD22566207
  • Cas Number : 1232365-42-2
  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3,3-difluoro-1-(hydroxymethyl)cyclobutyl)carbamate
Inchi Code 1S/C10H17F2NO3/c1-8(2,3)16-7(15)13-9(6-14)4-10(11,12)5-9/h14H,4-6H2,1-3H3,(H,13,15)
Inchi Key RNZVBWMKWVRXIY-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/Y54164
Availability details for
Quantity
  • Availability
  • Add to Cart