MAN517333469 Manchester Organics

Alogliptin benzoate

  • MDL number MFCD09833195
  • Cas Number : 850649-62-6
  • Molecular Weight : 461.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/Z54234
Availability details for
Quantity
  • Availability
  • Add to Cart