MAN517333482 Manchester Organics

Dicyclopropylmethanamine

  • MDL number MFCD06804453
  • Cas Number : 13375-29-6
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name dicyclopropylmethanamine
Inchi Code 1S/C7H13N/c8-7(5-1-2-5)6-3-4-6/h5-7H,1-4,8H2
Inchi Key GDGUATCKWWKTLM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/N54247
Availability details for
Quantity
  • Availability
  • Add to Cart