MAN517333554 Manchester Organics

tert-Butyl N-[3-(hydroxymethyl)oxetan-3-yl]carbamate

  • MDL number MFCD22415250
  • Cas Number : 1363382-11-9
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(hydroxymethyl)-3-oxetanylcarbamate
Inchi Code 1S/C9H17NO4/c1-8(2,3)14-7(12)10-9(4-11)5-13-6-9/h11H,4-6H2,1-3H3,(H,10,12)
Inchi Key IVFJORRHEURGIF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z54326
Availability details for
Quantity
  • Availability
  • Add to Cart