MAN640939750 Manchester Organics

[(2R)-1,4-Dioxan-2-yl]methanamine hydrochloride

  • MDL number MFCD27918519
  • Cas Number : 1523541-84-5
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/P54662
Availability details for
Quantity
  • Availability
  • Add to Cart