MAN640939959 Manchester Organics

3-(p-Tolyl)oxetan-3-amine hydrochloride

  • MDL number MFCD19381737
  • Molecular Weight : 199.67726

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R13991

Safety Information

MSDS https://www.manchesterorganics.com/R13991
Availability details for
Quantity
  • Availability
  • Add to Cart