MAN640940111 Manchester Organics

4-Benzyloxy-2,6-difluorobenzoic acid

  • MDL number MFCD09029982
  • Molecular Weight : 264.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B54995
Availability details for
Quantity
  • Availability
  • Add to Cart