MAN640940246 Manchester Organics

(5-Bromo-3-pyridyl)-morpholino-methanone

  • MDL number MFCD01788376
  • Molecular Weight : 271.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/M55143
Availability details for
Quantity
  • Availability
  • Add to Cart