MAN998716809 Manchester Organics

Perfluoro-2,4-dimethyl-3-ethylpentane : Perfluoro-2,4-dimethylheptane (approx. 70%:30%)

  • MDL number MFCD26405741
  • Molecular Weight : 868.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M49669
Availability details for
Quantity
  • Availability
  • Add to Cart