MAN998716842 Manchester Organics

3-Chloro-5-(trifluoromethoxy)phenol

  • MDL number MFCD09832330
  • Molecular Weight : 212.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.94
Country of Origin GB
Product Link https://www.manchesterorganics.com/M55557
Availability details for
Quantity
  • Availability
  • Add to Cart