MAN998716849 Manchester Organics

N-Methylpiperidin-4-amine dihydrochloride

  • MDL number MFCD04039797
  • Molecular Weight : 187.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E32044
Availability details for
Quantity
  • Availability
  • Add to Cart