MAN998716906 Manchester Organics

[(Z)-Octadec-9-enyl] methanesulfonate

  • MDL number MFCD00083434
  • Molecular Weight : 346.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A56167
Availability details for
Quantity
  • Availability
  • Add to Cart