MAN998716947 Manchester Organics

1-Methyl-5-phenyl-imidazol-2-amine

  • MDL number MFCD05668690
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/TA6390
Availability details for
Quantity
  • Availability
  • Add to Cart