MAN998717115 Manchester Organics

5-Norbornene-2-carboxaldehyde

  • MDL number MFCD00167568
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z55798
Availability details for
Quantity
  • Availability
  • Add to Cart