MAN998717151 Manchester Organics

(2-Benzyloxy-3-pyridyl)boronic acid

  • MDL number MFCD06798264
  • Molecular Weight : 229.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/D56101
Availability details for
Quantity
  • Availability
  • Add to Cart