MAN998717218 Manchester Organics

6-Bromo-3-methoxy-2-methyl-pyridine

  • MDL number MFCD11846419
  • Cas Number : 850882-87-0
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-3-methoxy-2-methylpyridine
Inchi Code 1S/C7H8BrNO/c1-5-6(10-2)3-4-7(8)9-5/h3-4H,1-2H3
Inchi Key HUBRRMYRFLDKED-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/X56417
Availability details for
Quantity
  • Availability
  • Add to Cart