MAN998717503 Manchester Organics

3-(4-(Trifluoromethyl)phenyl)oxetan-3-amine hydrochloride

  • MDL number MFCD20233322
  • Cas Number : 1349718-44-0
  • Molecular Weight : 253.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T13993
Availability details for
Quantity
  • Availability
  • Add to Cart