MAN998717835 Manchester Organics

Diethyl benzene-1,4-dicarboxylate

  • MDL number MFCD00039891
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T55884
Availability details for
Quantity
  • Availability
  • Add to Cart