MAN998717850 Manchester Organics

4-Hydrazinobenzenesulfonic acid hydrate

  • MDL number MFCD00460768
  • Molecular Weight : 206.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S55883
Availability details for
Quantity
  • Availability
  • Add to Cart