MAN998717857 Manchester Organics

2,6-Dichloro-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD09743917
  • Molecular Weight : 259.00942

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B56145

Safety Information

MSDS https://www.manchesterorganics.com/B56145
Availability details for
Quantity
  • Availability
  • Add to Cart